Order Entry
Export
Orders LinkContactUsLinkComponent
1-Tridecene 99%

1-Tridecene 99%

Bottle
Catalog # APOSOR95310-1G
Health
New Product
Supplier:   Apollo Scientific
1-Tridecene 99%
Catalog # APOSOR95310-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  2437-56-1
1-Tridecene 99%

Bottle

CAS Number:  2437-56-1
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Organic building block/reagent.

  • CAS Number:2437-56-1
  • Formula:C₁₃H₂₆
  • MDL Number:MFCD00008976
  • Molecular Weight:182,35 g/mol
  • EINECS:219-443-8
Specifications

Specifications

  • Pack type:
    Bottle

Specifications