Dimethylphenylsilane 97%
Bottle
Catalog # APOSOR72472-2.5KG
New Product
Supplier: Apollo Scientific
Dimethylphenylsilane 97%
Catalog # APOSOR72472-2.5KG
Supplier: Apollo Scientific
New Product
CAS Number: 766-77-8
Jump to:
Product Details & Documents
Usage: Organic building block/reagent.
Synonyms: Dimethyl(phenyl)silane
- CAS Number:766-77-8
- Formula:C₈H₁₂Si
- MDL Number:MFCD00008257
- Molecular Weight:136,27 g/mol
- EINECS:212-170-5
- UN:1993
Specifications
Specifications
- Pack type:Bottle



