Order Entry
Export
Orders LinkContactUsLinkComponent
Diphenylsilane 98%

Diphenylsilane 98%

Bottle
Catalog # APOSOR72471-1G
Irritant
New Product
Supplier:   Apollo Scientific
Diphenylsilane 98%
Catalog # APOSOR72471-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  775-12-2
Diphenylsilane 98%

Bottle

CAS Number:  775-12-2
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Organic building block/reagent

  • CAS Number:775-12-2
  • Formula:C₁₂H₁₂Si
  • MDL Number:MFCD00003002
  • Molecular Weight:184,31 g/mol
  • EINECS:212-271-4
Specifications

Specifications

  • Pack type:
    Bottle

Specifications