Order Entry
Export
Orders LinkContactUsLinkComponent
Calcium phosphate 95%

Calcium phosphate 95%

Bottle
Catalog # APOSIN1392-5G
Irritant
New Product
Supplier:   Apollo Scientific
Calcium phosphate 95%
Catalog # APOSIN1392-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  7758-87-4
Calcium phosphate 95%

Bottle

CAS Number:  7758-87-4
Price
21,80 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Inorganic salt or catalyst

Synonyms:  Calcium phosphate, Tricalcium orthophosphate, penta-Calcium hydroxide triphosphate, Calcium phosphate tribasic
  • CAS Number:7758-87-4
  • Formula:3Ca·2PO₄
  • MDL Number:MFCD00015984
  • Molecular Weight:310,17672 g/mol
  • EINECS:231-840-8
Specifications

Specifications

  • Pack type:
    Bottle

Specifications