Order Entry
Export
Orders LinkContactUsLinkComponent
Methyldiphenylsilane 99%

Methyldiphenylsilane 99%

Bottle
Catalog # APOSOR72475-500G
Flammable
Irritant
New Product
Supplier:   Apollo Scientific
Methyldiphenylsilane 99%
Catalog # APOSOR72475-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  776-76-1
Methyldiphenylsilane 99%

Bottle

CAS Number:  776-76-1
Price
642,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  Diphenylmethylsilane
  • CAS Number:776-76-1
  • Formula:C₁₃H₁₄Si
  • MDL Number:MFCD00008255
  • Molecular Weight:198,34 g/mol
  • EINECS:212-281-9
  • UN:1993
Specifications

Specifications

  • Pack type:
    Bottle
  • Cat. No.:
    APOSOR72475-500G

Specifications