Order Entry
Export
Orders LinkContactUsLinkComponent
 

4,7-DIHYDROXYCOUMARIN ≥98%

Bottle
Catalog # APOSBUP14724-500MG
New Product
Supplier:   Apollo Scientific
4,7-DIHYDROXYCOUMARIN ≥98%
Catalog # APOSBUP14724-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  1983-81-9
4,7-DIHYDROXYCOUMARIN ≥98%

Bottle

CAS Number:  1983-81-9
Price
164,00 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  4,7-Dihydroxy-2H-chromen-2-one
  • CAS Number:1983-81-9
  • Formula:C₉H₆O₄
  • Molecular Weight:178,14 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications