0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
4,7-Dihydroxycoumarin
BP0000733064
4,7-DIHYDROXYCOUMARIN
Bottle
APOSBUP14724-50MG
New Product
:
Apollo Scientific
4,7-DIHYDROXYCOUMARIN
APOSBUP14724-50MG
:
Apollo Scientific
New Product
:
4,7-DIHYDROXYCOUMARIN
Bottle
:
44,90 €
1 * 50 mg
-
+
Pack type:
Bottle