Order Entry
Export
Orders LinkContactUsLinkComponent
 

Umbelliferone ≥98%

Bottle
  APOSBUP14594-10G
New Product
Umbelliferone ≥98%
  APOSBUP14594-10G
New Product
 :  93-35-6
Umbelliferone ≥98%

Bottle

 :  93-35-6
 
529,00 €
   
1 * 10 g
 

Usage: Biochemical/life science applications

 :  Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone
  •  :93-35-6
  •  :C₉H₆O₃
  •  :162,14 g/mol
 

 

  • Pack type:
    Bottle