Order Entry
Export
Orders LinkContactUsLinkComponent
Urea ≥95%

Urea ≥95%

Bottle
Catalog # APOSBUP14678-100MG
New Product
Supplier:   Apollo Scientific
Urea ≥95%
Catalog # APOSBUP14678-100MG
Supplier:  Apollo Scientific
New Product
CAS Number:  57-13-6
Urea ≥95%

Bottle

CAS Number:  57-13-6
Price
53,60 €
Log in for availability
1 * 100 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
  • CAS Number:57-13-6
  • Formula:CH₄N₂O
  • Molecular Weight:60,06 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications