Order Entry
Export
Orders LinkContactUsLinkComponent
Umbelliferone

Umbelliferone

Bottle
Catalog # APOSBUP14594-5G
New Product
Supplier:   Apollo Scientific
Umbelliferone
Catalog # APOSBUP14594-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  
Umbelliferone

Bottle

CAS Number:  
Price
348,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone
  • Molecular Weight:162,14 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications