Order Entry
Export
Orders LinkContactUsLinkComponent
Umbelliferone ≥98%

Umbelliferone ≥98%

Bottle
Catalog # APOSBUP14594-50MG
New Product
Supplier:   Apollo Scientific
Umbelliferone ≥98%
Catalog # APOSBUP14594-50MG
Supplier:  Apollo Scientific
New Product
CAS Number:  93-35-6
Umbelliferone ≥98%

Bottle

CAS Number:  93-35-6
Price
39,10 €
Log in for availability
1 * 50 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone
  • CAS Number:93-35-6
  • Formula:C₉H₆O₃
  • Molecular Weight:162,14 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications