Order Entry
Export
Orders LinkContactUsLinkComponent
3-Bromopentane 99%

3-Bromopentane 99%

Bottle
Catalog # APOSOR95211-500G
Flammable
Irritant
New Product
Supplier:   Apollo Scientific
3-Bromopentane 99%
Catalog # APOSOR95211-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  1809-10-5
3-Bromopentane 99%

Bottle

CAS Number:  1809-10-5
Price
1 160,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Organic building block/reagent.

  • CAS Number:1809-10-5
  • Formula:C₅H₁₁Br
  • MDL Number:MFCD00000158
  • Molecular Weight:151,04 g/mol
  • EINECS:217-314-0
  • UN:1993
Specifications

Specifications

  • Pack type:
    Bottle

Specifications