3-Bromopentane 99%
Bottle
Catalog # APOSOR95211-1G
New Product
Supplier: Apollo Scientific
3-Bromopentane 99%
Catalog # APOSOR95211-1G
Supplier: Apollo Scientific
New Product
CAS Number: 1809-10-5
Jump to:
Product Details & Documents
Usage: Organic building block/reagent.
- CAS Number:1809-10-5
- Formula:C₅H₁₁Br
- MDL Number:MFCD00000158
- Molecular Weight:151,04 g/mol
- EINECS:217-314-0
- UN:1993
Specifications
Specifications
- Pack type:Bottle



