Order Entry
Export
Orders LinkContactUsLinkComponent
2,2'-Biquinoline 99%

2,2'-Biquinoline 99%

Bottle
Catalog # APOSOR74143-100G
New Product
Supplier:   Apollo Scientific
2,2'-Biquinoline 99%
Catalog # APOSOR74143-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  119-91-5
2,2'-Biquinoline 99%

Bottle

CAS Number:  119-91-5
Price
393,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:119-91-5
  • Formula:C₁₈H₁₂N₂
  • MDL Number:MFCD00006740
  • Molecular Weight:256,308 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications