Order Entry
Export
Orders LinkContactUsLinkComponent
1,7-Dibromoheptane 95%

1,7-Dibromoheptane 95%

Bottle
Catalog # APOSOR1556-500G
Toxic
New Product
Supplier:   Apollo Scientific
1,7-Dibromoheptane 95%
Catalog # APOSOR1556-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  4549-31-9
1,7-Dibromoheptane 95%

Bottle

CAS Number:  4549-31-9
Price
1 150,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Heptamethylene dibromide
  • CAS Number:4549-31-9
  • Formula:BrH₂C(CH₂)₅CH₂Br
  • MDL Number:MFCD00000274
  • Molecular Weight:258 g/mol
  • Boiling Pt:255 °C
  • Density:1.51 g/cm³ (25 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:224-910-4
  • UN:2810
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle

Specifications