Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
17-DMAG
17-DMAG
17-DMAG
17-DMAG
Bottle
APOSBID1002-500MG
New Product
:
Apollo Scientific
17-DMAG
APOSBID1002-500MG
:
Apollo Scientific
New Product
:
467214-20-6
Expand image
17-DMAG
Bottle
Add to cart
:
467214-20-6
2 750,00 €
Unit of Measure
1 * 500 mg
-
+
Add to cart
:
17-Dimethylaminoethylamino-17-demethoxygeldanamycin
:
467214-20-6
:
C₃₂H₄₈N₄O₈
:
MFCD08457919
:
616.8 g/mol
:
Freezer
Pack type:
Bottle
Cat. No.:
APOSBID1002-500MG