Order Entry
Export
Orders LinkContactUsLinkComponent
2,5-Dihydroxyterephthalaldehyde

2,5-Dihydroxyterephthalaldehyde

Bottle
Catalog # APOSOR315542-5G
New Product
Supplier:   Apollo Scientific
2,5-Dihydroxyterephthalaldehyde
Catalog # APOSOR315542-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  1951-36-6
2,5-Dihydroxyterephthalaldehyde

Bottle

CAS Number:  1951-36-6
Price
268,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:1951-36-6
  • Formula:C₈H₆O₄
  • MDL Number:MFCD22055335
  • Molecular Weight:166.131 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications