2,5-Dihydroxyterephthalaldehyde
Bottle
Catalog # APOSOR315542-100MG
New Product
Supplier: Apollo Scientific
2,5-Dihydroxyterephthalaldehyde
Catalog # APOSOR315542-100MG
Supplier: Apollo Scientific
New Product
CAS Number: 1951-36-6
Jump to:
Product Details & Documents
- CAS Number:1951-36-6
- Formula:C₈H₆O₄
- MDL Number:MFCD22055335
- Molecular Weight:166.131 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle



