Order Entry
Export
Orders LinkContactUsLinkComponent
Propidium Iodide

Propidium Iodide

Bottle
Catalog # APOSBIP4355-25MG
Irritant
Health
New Product
Supplier:   Apollo Scientific
Propidium Iodide
Catalog # APOSBIP4355-25MG
Supplier:  Apollo Scientific
New Product
CAS Number:  25535-16-4
Propidium Iodide

Bottle

CAS Number:  25535-16-4
Price
99,90 €
Log in for availability
1 * 25 mg
Product Details & Documents

Fluorescent staining compound.

Synonyms:  3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide
  • CAS Number:25535-16-4
  • Formula:C₂₇H₃₄I₂N₄
  • MDL Number:MFCD00011921
  • Molecular Weight:668.4 g/mol
  • Melting Pt:220…225 °C (dec.)
  • Storage Temperature:Refrigerator
  • EINECS:247-081-0
Specifications

Specifications

  • Pack type:
    Bottle

Specifications