Order Entry
Export
Orders LinkContactUsLinkComponent
2,6-Dichloronicotinamide 95%

2,6-Dichloronicotinamide 95%

Bottle
Catalog # APOSOR942378-500G
New Product
Supplier:   Apollo Scientific
2,6-Dichloronicotinamide 95%
Catalog # APOSOR942378-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  62068-78-4
2,6-Dichloronicotinamide 95%

Bottle

CAS Number:  62068-78-4
Price
1 150,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  2,6-Dichloropyridine-3-carboxamide
  • CAS Number:62068-78-4
  • Formula:C₆H₄Cl₂N₂O
  • MDL Number:MFCD03230010
  • Molecular Weight:191.015 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications