Order Entry
Export
Orders LinkContactUsLinkComponent
2,4-Hexanedione 97%

2,4-Hexanedione 97%

Bottle
Catalog # APOSOR938383-100G
New Product
Supplier:   Apollo Scientific
2,4-Hexanedione 97%
Catalog # APOSOR938383-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  3002-24-2
2,4-Hexanedione 97%

Bottle

CAS Number:  3002-24-2
Price
828,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:3002-24-2
  • Formula:C₆H₁₀O₂
  • MDL Number:MFCD00009318
  • Molecular Weight:114.14 g/mol
  •  :Density: 0.9538 g/cm³ (15 °C)
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications