Order Entry
Export
Orders LinkContactUsLinkComponent
Pyridine-2,6-dicarbonitrile 95%

Pyridine-2,6-dicarbonitrile 95%

Bottle
Catalog # APOSOR938560-100G
New Product
Supplier:   Apollo Scientific
Pyridine-2,6-dicarbonitrile 95%
Catalog # APOSOR938560-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  2893-33-6
Pyridine-2,6-dicarbonitrile 95%

Bottle

CAS Number:  2893-33-6
Price
183,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  2,6-Dicyanopyridine
  • CAS Number:2893-33-6
  • Formula:C₇H₃N₃
  • MDL Number:MFCD00129021
  • Molecular Weight:129.12 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications