Order Entry
Export
Orders LinkContactUsLinkComponent
1,5-Pentanediol 97%

1,5-Pentanediol 97%

Bottle
Catalog # APOSOR921506-25G
New Product
Supplier:   Apollo Scientific
1,5-Pentanediol 97%
Catalog # APOSOR921506-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  111-29-5
1,5-Pentanediol 97%

Bottle

CAS Number:  111-29-5
Price
21,80 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Pentane-1,5-diol, 1,5-Dihydroxy pentane
  • CAS Number:111-29-5
  • Formula:C₅H₁₂O₂
  • MDL Number:MFCD00002978
  • Molecular Weight:104.15 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications