Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Diethoxybenzene 97%

1,3-Diethoxybenzene 97%

Bottle
Catalog # APOSOR920967-2.5KG
New Product
Supplier:   Apollo Scientific
1,3-Diethoxybenzene 97%
Catalog # APOSOR920967-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  2049-73-2
1,3-Diethoxybenzene 97%

Bottle

CAS Number:  2049-73-2
Price
2 280,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
  • CAS Number:2049-73-2
  • Formula:C₁₀H₁₄O₂
  • MDL Number:MFCD00015145
  • Molecular Weight:166.22 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications