Order Entry
Export
Orders LinkContactUsLinkComponent
 

(R,R)-(-)-3,6-Octanediol 96%

Bottle
Catalog # APOSOR932401-5G
New Product
Supplier:   Apollo Scientific
(R,R)-(-)-3,6-Octanediol 96%
Catalog # APOSOR932401-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  129619-37-0
(R,R)-(-)-3,6-Octanediol 96%

Bottle

CAS Number:  129619-37-0
Price
508,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  (3R,6R)-Octane-3,6-diol, (R,R)-(-)-3,6-Dihydroxy octane, (3R,6R)-(-)-3,6-Octanediol
  • CAS Number:129619-37-0
  • Formula:C₈H₁₈O₂
  • MDL Number:MFCD01631087
  • Molecular Weight:146.23 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications