iFSP1 ≥98%
Bottle
Catalog # APOSBUP10939-500MG
New Product
Supplier: Apollo Scientific
iFSP1 ≥98%
Catalog # APOSBUP10939-500MG
Supplier: Apollo Scientific
New Product
CAS Number: 150651-39-1
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications
- Store at 2 to 8 °C
- Non-hazardous
Synonyms: 1-amino-3-(4-methylphenyl)-pyrido[1,2-a]benzimidazole-2,4-dicarbonitrile, 1-Amino-3-p-tolyl-benzo[4,5]imidazo[1,2-a]pyridine-2,4-dicarbonitrile
- CAS Number:150651-39-1
- Formula:C₂₀H₁₃N₅
- MDL Number:MFCD01572665
- Molecular Weight:323,35 g/mol
- Storage Temperature:Refrigerator
- EINECS:851-482-1
Specifications
Specifications
- Pack type:Bottle



