Order Entry
Export
Orders LinkContactUsLinkComponent
iFSP1 ≥98%

iFSP1 ≥98%

Bottle
  APOSBUP10939-100MG
New Product
iFSP1 ≥98%
  APOSBUP10939-100MG
New Product
 :  150651-39-1
iFSP1 ≥98%

Bottle

 :  150651-39-1
 
417,00 €
Log in for availability
1 * 100 mg
 

Usage: Biochemical/life science applications

  • Store at 2 to 8 °C
  • Non-hazardous
Synonyms:  1-amino-3-(4-methylphenyl)-pyrido[1,2-a]benzimidazole-2,4-dicarbonitrile, 1-Amino-3-p-tolyl-benzo[4,5]imidazo[1,2-a]pyridine-2,4-dicarbonitrile
  • CAS Number:150651-39-1
  • Formula:C₂₀H₁₃N₅
  • MDL Number:MFCD01572665
  • Molecular Weight:323,35 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:851-482-1
Specifications

Specifications

  • Pack type:
    Bottle

Specifications