Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
iFSP1
BP0000728750
iFSP1
Print
iFSP1
Bottle
Catalog # APOSBUP10939-25MG
New Product
Supplier:
Apollo Scientific
iFSP1
Catalog # APOSBUP10939-25MG
Supplier:
Apollo Scientific
New Product
CAS Number:
Expand image
Return to Top
iFSP1
Bottle
Specifications
Add to cart
Jump to:
Specifications
CAS Number:
Price
122,00 €
Log in
for availability
Unit of Measure
1 * 25 mg
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Specifications
Pack type:
Bottle
Specifications