Order Entry
Export
Orders LinkContactUsLinkComponent
Tobramycin ≥98%

Tobramycin ≥98%

Bottle
Catalog # APOSBUP15965-50MG
New Product
Supplier:   Apollo Scientific
Tobramycin ≥98%
Catalog # APOSBUP15965-50MG
Supplier:  Apollo Scientific
New Product
CAS Number:  32986-56-4
Tobramycin ≥98%

Bottle

CAS Number:  32986-56-4
Price
42,00 €
Log in for availability
1 * 50 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  (2S,3R,4S,5S,6R)-4-Amino-2-(((1S,2S,3R,4S,6R)-4,6-diamino-3-(((2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxytetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,5-diol
  • CAS Number:32986-56-4
  • Formula:C₁₈H₃₇N₅O₉
  • Molecular Weight:467,51 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications