0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
D(+)-Galactose
BP0000707512
D-Galactose
Bottle
APOSBUP17773-1G
New Product
:
Apollo Scientific
D-Galactose
APOSBUP17773-1G
:
Apollo Scientific
New Product
:
D-Galactose
Bottle
:
63,70 €
1 * 1 g
-
+
Pack type:
Bottle