Order Entry
Export
Orders LinkContactUsLinkComponent
8-Hydroxyquinoline ≥98%

8-Hydroxyquinoline ≥98%

Bottle
Catalog # APOSBUP18706-10G
New Product
Supplier:   Apollo Scientific
8-Hydroxyquinoline ≥98%
Catalog # APOSBUP18706-10G
Supplier:  Apollo Scientific
New Product
CAS Number:  148-24-3
8-Hydroxyquinoline ≥98%

Bottle

CAS Number:  148-24-3
Price
86,90 €
Log in for availability
1 * 10 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  8-Hydroxyquinoline
  • CAS Number:148-24-3
  • Formula:C₉H₇NO
  • Molecular Weight:145,16 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications