Order Entry
Export
Orders LinkContactUsLinkComponent
 

Sarcosine ≥98%

Bottle
  APOSBUP18651-10G
New Product
Sarcosine ≥98%
  APOSBUP18651-10G
New Product
 :  107-97-1
Sarcosine ≥98%

Bottle

 :  107-97-1
 
252,00 €
   
1 * 10 g
 

Usage: Biochemical/life science applications.

 :  H-Sar-OH, N-Methylglycine, Methylaminoacetic acid
  •  :107-97-1
  •  :C₃H₇NO₂
  •  :89,09 g/mol
 

 

  • Pack type:
    Bottle