Order Entry
Export
Orders LinkContactUsLinkComponent
 

Sarcosine ≥98%

Bottle
Catalog # APOSBUP18651-1G
New Product
Supplier:   Apollo Scientific
Sarcosine ≥98%
Catalog # APOSBUP18651-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  107-97-1
Sarcosine ≥98%

Bottle

CAS Number:  107-97-1
Price
71,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  H-Sar-OH, N-Methylglycine, Methylaminoacetic acid
  • CAS Number:107-97-1
  • Formula:C₃H₇NO₂
  • Molecular Weight:89,09 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications