Chloramphenicol ≥98%
Bottle
Catalog # APOSBUP18539-10G
New Product
Supplier: Apollo Scientific
Chloramphenicol ≥98%
Catalog # APOSBUP18539-10G
Supplier: Apollo Scientific
New Product
CAS Number: 56-75-7
Jump to:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol
- CAS Number:56-75-7
- Formula:C₁₁H₁₂Cl₂N₂O₅
- Molecular Weight:323,13 g/mol
Specifications
Specifications
- Pack type:Bottle



