Order Entry
Export
Orders LinkContactUsLinkComponent
 

Chloramphenicol ≥98%

Bottle
Catalog # APOSBUP18539-1G
New Product
Supplier:   Apollo Scientific
Chloramphenicol ≥98%
Catalog # APOSBUP18539-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  56-75-7
Chloramphenicol ≥98%

Bottle

CAS Number:  56-75-7
Price
62,30 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol
  • CAS Number:56-75-7
  • Formula:C₁₁H₁₂Cl₂N₂O₅
  • Molecular Weight:323,13 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications