Order Entry
Export
Orders LinkContactUsLinkComponent
Hygromycin B ≥98%

Hygromycin B ≥98%

Bottle
Catalog # APOSBUP17695-500MG
New Product
Supplier:   Apollo Scientific
Hygromycin B ≥98%
Catalog # APOSBUP17695-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  31282-04-9
Hygromycin B ≥98%

Bottle

CAS Number:  31282-04-9
Price
89,80 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol
  • CAS Number:31282-04-9
  • Formula:C₂₀H₃₇N₃O₁₃
  • Molecular Weight:527,52 g/mol
Specifications

Specifications

  • Pack type:
    Bottle
  • Cat. No.:
    APOSBUP17695-500MG

Specifications