Order Entry
Export
Orders LinkContactUsLinkComponent
Biotin ≥98%

Biotin ≥98%

Bottle
Catalog # APOSBUP18500-500MG
New Product
Supplier:   Apollo Scientific
Biotin ≥98%
Catalog # APOSBUP18500-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  58-85-5
Biotin ≥98%

Bottle

CAS Number:  58-85-5
Price
59,40 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

  • Store at 2 to 8 °C Non-hazardous
Synonyms:  Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
  • CAS Number:58-85-5
  • Formula:C₁₀H₁₆N₂O₃S
  • Molecular Weight:244,31 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:200-399-3
Specifications

Specifications

  • Pack type:
    Bottle

Specifications