Orders LinkContactUsLinkComponent
SN-38 ≥98%
Bottle
  APOSBUP14103-100MG
New Product
SN-38 ≥98%
  APOSBUP14103-100MG
New Product
 :  86639-52-3
SN-38 ≥98%
Bottle
 :  86639-52-3
98,40 €
1 * 100 mg
Usage: Biochemical/life science applications
 :  10-hydroxy-7-ethylcamptothecin, (S)-4,11-Diethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 7-Ethyl-10-hydroxy-20(S)-Camptothecin, SN-38, NK 012
  •  :86639-52-3
  •  :C₂₂H₂₀N₂O₅
  •  :392,4 g/mol
  • Pack type:
    Bottle