SN-38 ≥98%
Bottle
Catalog # APOSBUP14103-50MG
New Product
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications
Synonyms: 10-hydroxy-7-ethylcamptothecin, (S)-4,11-Diethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 7-Ethyl-10-hydroxy-20(S)-Camptothecin, SN-38, NK 012
- CAS Number:86639-52-3
- Formula:C₂₂H₂₀N₂O₅
- Molecular Weight:392,4 g/mol
Specifications
Specifications
- Pack type:Bottle
- Cat. No.:APOSBUP14103-50MG



