Order Entry
Export
Orders LinkContactUsLinkComponent
SN-38 ≥98%

SN-38 ≥98%

Bottle
Catalog # APOSBUP14103-25MG
New Product
Supplier:   Apollo Scientific
SN-38 ≥98%
Catalog # APOSBUP14103-25MG
Supplier:  Apollo Scientific
New Product
CAS Number:  86639-52-3
SN-38 ≥98%

Bottle

CAS Number:  86639-52-3
Price
50,70 €
Log in for availability
1 * 25 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  10-hydroxy-7-ethylcamptothecin, (S)-4,11-Diethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 7-Ethyl-10-hydroxy-20(S)-Camptothecin, SN-38, NK 012
  • CAS Number:86639-52-3
  • Formula:C₂₂H₂₀N₂O₅
  • Molecular Weight:392,4 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications