Order Entry
Export
Orders LinkContactUsLinkComponent
AGI-6780 ≥95%

AGI-6780 ≥95%

Bottle
Catalog # APOSBUP09554-50MG
New Product
Supplier:   Apollo Scientific
AGI-6780 ≥95%
Catalog # APOSBUP09554-50MG
Supplier:  Apollo Scientific
New Product
CAS Number:  1432660-47-3
AGI-6780 ≥95%

Bottle

CAS Number:  1432660-47-3
Price
526,00 €
Log in for availability
1 * 50 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  N-Cyclopropyl-4-(thiophen-3-yl)-3-(3-(3-(trifluoromethyl)phenyl)ureido)benzenesulphonamide
  • CAS Number:1432660-47-3
  • Formula:C₂₁H₁₈F₃N₃O₃S₂
  • Molecular Weight:481,51 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications