Order Entry
Export
Orders LinkContactUsLinkComponent
beta-Citronellol 95%

beta-Citronellol 95%

Bottle
Catalog # APOSOR6011-1KG
Irritant
New Product
Supplier:   Apollo Scientific
beta-Citronellol 95%
Catalog # APOSOR6011-1KG
Supplier:  Apollo Scientific
New Product
CAS Number:  106-22-9
beta-Citronellol 95%

Bottle

CAS Number:  106-22-9
Price
158,00 €
Log in for availability
1 * 1 kg
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  DL-Citronellol, (±)-Citronellol, Citronellol, β-Citronellol, 3,7-Dimethyloct-6-en-1-ol
  • CAS Number:106-22-9
  • Formula:C₁₀H₂₀O
  • MDL Number:MFCD00002935
  • Molecular Weight:156,2652 g/mol
  • EINECS:203-375-0
Specifications

Specifications

  • Pack type:
    Bottle

Specifications