Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Propanedithiol 97%

1,3-Propanedithiol 97%

Bottle
Catalog # APOSOR70117-2.5KG
New Product
Supplier:   Apollo Scientific
1,3-Propanedithiol 97%
Catalog # APOSOR70117-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  109-80-8
1,3-Propanedithiol 97%

Bottle

CAS Number:  109-80-8
Price
1 660,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  1,3-Dimercaptopropane
  • CAS Number:109-80-8
  • Formula:C₃H₈S₂
  • MDL Number:MFCD00004904
  • Molecular Weight:108.23 g/mol
  • Boiling Pt:168…170 °C
  • Melting Pt:–79 °C
  • Density:1.077 g/cm³ (25 °C)
  • Flash Pt:63 °C
  • Storage Temperature:Refrigerator
  • UN:3334
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Bottle

Specifications