Phenacetin ≥98%
Bottle
Catalog # APOSBUP18451-1G
New Product
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications
Synonyms: Acetphenetidin, Acetophenetidin, Phenacetin, N-(4-Ethoxyphenyl)acetamide, 4-Acetophenetidide
- CAS Number:62-44-2
- Formula:C₁₀H₁₃NO₂
- Molecular Weight:179,22 g/mol
- EINECS:200-533-0
Specifications
Specifications
- Pack type:Bottle



