Order Entry
Export
Orders LinkContactUsLinkComponent
Docosanoic acid ≥98%

Docosanoic acid ≥98%

Bottle
Catalog # APOSBUP18445-500MG
New Product
Supplier:   Apollo Scientific
Docosanoic acid ≥98%
Catalog # APOSBUP18445-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  112-85-6
Docosanoic acid ≥98%

Bottle

CAS Number:  112-85-6
Price
62,30 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  Docosanoic acid
  • CAS Number:112-85-6
  • Formula:C₂₂H₄₄O₂
  • Molecular Weight:340,58 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications