17-ODYA ≥98%
Bottle
Catalog # APOSBUP04765-100MG
New Product
Supplier: Apollo Scientific
17-ODYA ≥98%
Catalog # APOSBUP04765-100MG
Supplier: Apollo Scientific
New Product
CAS Number: 34450-18-5
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications
- CAS Number:34450-18-5
- Formula:C₁₈H₃₂O₂
- Molecular Weight:280,45 g/mol
Specifications
Specifications
- Pack type:Bottle



