Order Entry
Export
Orders LinkContactUsLinkComponent
17-ODYA ≥98%

17-ODYA ≥98%

Bottle
Catalog # APOSBUP04765-25MG
New Product
Supplier:   Apollo Scientific
17-ODYA ≥98%
Catalog # APOSBUP04765-25MG
Supplier:  Apollo Scientific
New Product
CAS Number:  34450-18-5
17-ODYA ≥98%

Bottle

CAS Number:  34450-18-5
Price
346,00 €
Log in for availability
1 * 25 mg
Product Details & Documents

Usage: Biochemical/life science applications

  • CAS Number:34450-18-5
  • Formula:C₁₈H₃₂O₂
  • Molecular Weight:280,45 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications