Order Entry
Export
Orders LinkContactUsLinkComponent
Dodecanedioic acid ≥98%

Dodecanedioic acid ≥98%

Bottle
Catalog # APOSBUP19772-5G
New Product
Supplier:   Apollo Scientific
Dodecanedioic acid ≥98%
Catalog # APOSBUP19772-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  693-23-2
Dodecanedioic acid ≥98%

Bottle

CAS Number:  693-23-2
Price
69,50 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Dodecanedioic acid, 1,10-Decanedicarboxylic acid
  • CAS Number:693-23-2
  • Formula:C₁₂H₂₂O₄
  • Molecular Weight:230,3 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications