0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
FGIN-1-43
BP0000724366
FGIN-1-43
Bottle
APOSBUP05796-50MG
New Product
:
Apollo Scientific
FGIN-1-43
APOSBUP05796-50MG
:
Apollo Scientific
New Product
:
FGIN-1-43
Bottle
:
354,00 €
1 * 50 mg
-
+
Pack type:
Bottle