Order Entry
Export
Orders LinkContactUsLinkComponent
Palmitic acid ≥98%

Palmitic acid ≥98%

Bottle
Catalog # APOSBUP19708-5G
New Product
Supplier:   Apollo Scientific
Palmitic acid ≥98%
Catalog # APOSBUP19708-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  57-10-3
Palmitic acid ≥98%

Bottle

CAS Number:  57-10-3
Price
62,30 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Hexadecanoic acid, Hexadecylic acid
  • CAS Number:57-10-3
  • Formula:C₁₆H₃₂O₂
  • Molecular Weight:256,42 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications